Gene Transcripts

Gene Symbol
Stau1
Entrez Gene ID
20853
Gene Name
staufen (RNA binding protein) homolog 1 (Drosophila)
Gene Aliases
5830401L18Rik, AW549911, C85792, Stau
Chromosome Location
Chr. 2: 166947549 - 166996299 on Build GRCm38
UniGene ID
Mm.73276
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109905.2 NP_001103375.1 11 1706
NM_001109906.2 NP_001103376.1 11 1724
NM_001291160.1 NP_001278089.1 11 1706
NM_011490.4 NP_035620.1 11 1700
GenBank mRNA AF061942.1 - 11 1700
AF395842.1 - 10 1427
AK005002.2 - 11 1647
AK139532.1 - 11 1653
AK145826.1 - 11 1691
AK151835.1 - 11 1512
AK152269.1 - 11 1518
AK152744.1 - 11 1512
AK158952.1 - 6 2287
AK159602.1 - 11 1641
AK159781.1 - 11 1676
AK160979.1 - 11 1665
AK167637.1 - 11 1647
AK177429.1 - 2 66
AK179726.1 - 3 205
AY391773.1 - 11 1372
BC012959.1 - 11 1670 4188394
BC082277.1 - 11 1643

View Full Product Details