Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000207.2 | NP_000198.1 | 3 | 344 | |
NM_001185097.1 | NP_001172026.1 | 3 | 370 | ||
NM_001185098.1 | NP_001172027.1 | 2 | 523 | ||
NM_001291897.1 | NP_001278826.1 | 3 | 404 | ||
GenBank mRNA | AY899304.1 | - | 3 | 340 | |
BC005255.1 | - | 3 | 344 | 3950204 | |
BM510347.1 | - | 3 | 365 | ||
BM510748.1 | - | 3 | 348 | ||
BP322143.1 | - | 2 | 523 | ||
BT006808.1 | - | 2 | 285 | ||
DQ778082.1 | - | 2 | 249 | ||
JF909299.1 | - | 2 | 238 | ||
JQ951950.1 | - | 3 | 329 | ||
X70508.1 | - | 3 | 329 |