Gene Transcripts

Gene Symbol
Morf4l1
Entrez Gene ID
21761
Gene Name
mortality factor 4 like 1
Gene Aliases
MORFRG15, MRG15, TEG-189, Tex189, mKIAA4002
Chromosome Location
Chr. 9: 90091669 - 90114820 on Build GRCm38
UniGene ID
Mm.426209
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039147.2 NP_001034236.1 8 786
NM_024431.3 NP_077751.1 7 669
GenBank mRNA AB042241.1 - 7 518
AF319620.1 - 7 517
AF319621.1 - 8 634
AK010528.1 - 7 438
AK011249.1 - 7 546
AK011441.1 - 7 441
AK016414.1 - 7 636
AK079115.1 - 3 119
AK146210.1 - 7 504
AK146785.1 - 6 399
AK150925.1 - 7 470
AK153545.1 - 2 102
AK167079.1 - 7 428
AK168327.1 - 7 504
AK168585.1 - 7 449
AK168608.1 - 7 451
AK168674.1 - 7 500
AK169065.1 - 7 501
AK220441.1 - 5 3445
BC003894.1 - 7 521 3593982
BC017619.1 - 7 521 4923923
BC083094.1 - 7 454
BC083118.1 - 7 530 5371036
BC085103.1 - 8 636 6490291
BC092293.1 - 7 533 5068516
BC103783.1 - 7 533 6529024

View Full Product Details