Gene Transcripts

Gene Symbol
Tpbpa
Entrez Gene ID
21984
Gene Name
trophoblast specific protein alpha
Gene Aliases
4311, AV033951, Tb-1, Tb1, Tpbp, b2b1247Clo
Chromosome Location
Chr. 13: 60938692 - 60941935 on Build GRCm38
UniGene ID
Mm.2372
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009411.4 NP_033437.2 3 241
GenBank mRNA AK005406.1 - 3 241
AK005417.1 - 3 242
AK005438.1 - 3 241
AK005446.1 - 3 242
AK005454.1 - 3 243
AK005499.1 - 3 242
AK011401.1 - 3 242
AK014443.1 - 3 243
AK018797.1 - 3 242
AK018800.1 - 3 241
AK018802.1 - 3 241
AK018803.1 - 3 242
AK018806.1 - 3 242
AK018807.1 - 3 242
AK018811.1 - 3 242
AK018812.1 - 3 242
AK018813.1 - 3 242
AK018814.1 - 3 242
AK018815.1 - 3 241
AK018819.1 - 3 242
AK018821.1 - 3 242
AK018822.1 - 3 243
AK018823.1 - 3 242
AK018825.1 - 3 243
AK018826.1 - 3 242
AK028071.1 - 3 241
AK160140.1 - 3 235
AY034575.1 - 3 217
BC099391.1 - 3 228 30787380
X17071.1 - 3 233

View Full Product Details