Gene Transcripts

Gene Symbol
Zp2
Entrez Gene ID
22787
Gene Name
zona pellucida glycoprotein 2
Gene Aliases
Zp-2
Chromosome Location
Chr. 7: 120126772 - 120145754 on Build GRCm38
UniGene ID
Mm.6510
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011775.6 NP_035905.1 5 397
GenBank mRNA AK135903.1 - 5 397
AK138699.1 - 5 398
BC071183.1 - 5 379 30473497
M34148.1 - 5 397

View Full Product Details