Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001267040.1 | NP_001253969.1 | 2 | 146 | |
NM_004973.3 | NP_004964.2 | 2 | 646 | ||
XM_005249089.3 | XP_005249146.1 | 2 | 126 | ||
XM_011514579.2 | XP_011512881.1 | 4 | 790 | ||
XM_017010833.1 | XP_016866322.1 | 4 | 790 | ||
XM_017010834.1 | XP_016866323.1 | 2 | 646 | ||
GenBank mRNA | AK292861.1 | - | 2 | 492 | |
AK302624.1 | - | 2 | 141 | ||
AK303610.1 | - | 2 | 89 | ||
BC046246.1 | - | 2 | 363 | 4520786 | |
U57592.1 | - | 2 | 325 |