Gene Transcripts

Gene Symbol
Osr1
Entrez Gene ID
23967
Gene Name
odd-skipped related 1 (Drosophila)
Gene Aliases
Odd1, Osr
Chromosome Location
Chr. 12: 9574442 - 9581500 on Build GRCm38
UniGene ID
Mm.45055
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011859.3 NP_035989.1 2 895
GenBank mRNA AF117814.1 - 1 597
BC023922.1 - 2 851 5323948

View Full Product Details