Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011894.2 | NP_036024.2 | 7 | 942 | |
GenBank mRNA | AB016835.1 | - | 7 | 770 | |
AK043375.1 | - | 987 | |||
AK161427.1 | - | 6 | 924 | ||
AK165033.1 | - | 7 | 977 | ||
BC018237.1 | - | 7 | 974 | 3482670 | |
BC053741.1 | - | 7 | 942 | 6336511 |