Gene Transcripts

Gene Symbol
Aldh1a7
Entrez Gene ID
26358
Gene Name
aldehyde dehydrogenase family 1, subfamily A7
Gene Aliases
AI987940, Ahd2-like, Aldh-pb, Aldh1a4, Aldhpb
Chromosome Location
Chr. 19: 20692953 - 20727556 on Build GRCm38
UniGene ID
Mm.14609
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011921.2 NP_036051.1 6 563
GenBank mRNA BC032880.1 - 6 540
BC046315.1 - 6 537 6314273
BC139412.1 - 6 540 8862434
BC139413.1 - 6 540 8862435
U96401.1 - 6 563

View Full Product Details