Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011971.4 | NP_036101.1 | 5 | 543 | |
GenBank mRNA | AF060092.1 | - | 5 | 505 | |
AK002331.1 | - | 5 | 544 | ||
AK010488.1 | - | 5 | 524 | ||
AK153162.1 | - | 5 | 576 | ||
BC014783.1 | - | 5 | 510 | 3500033 | |
BC098233.1 | - | 4 | 505 | 1547611 |