Gene Transcripts

Gene Symbol
Hunk
Entrez Gene ID
26559
Gene Name
hormonally upregulated Neu-associated kinase
Gene Aliases
Bstk1, Mak-v
Chromosome Location
Chr. 16: 90385797 - 90499555 on Build GRCm38
UniGene ID
Mm.125874
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015755.2 NP_056570.1 2 333
GenBank mRNA AF055919.2 - 2 333
AF167987.1 - 2 345
AK144639.1 - 2 316
AK147425.1 - 2 718
AK147734.1 - 2 87
BC137788.1 - 2 394 8860807
BC137790.1 - 2 393 8860809

View Full Product Details