Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026794.2 | NP_081070.1 | 3 | 179 | |
GenBank mRNA | AK003863.1 | - | 3 | 187 | |
AK049702.1 | - | 2 | 2070 | ||
AK078209.1 | - | 2 | 291 | ||
AK085962.1 | - | 1 | 2119 | ||
AK213061.1 | - | 2 | 69 | ||
BC027548.1 | - | 3 | 179 | 1532650 |