Gene Transcripts
- Gene Symbol
- Dguok
- Entrez Gene ID
- 27369
- Gene Name
- deoxyguanosine kinase
- Gene Aliases
- dGK, dGK1, dGK2, dGK3
- Chromosome Location
- Chr. 6: 83480214 - 83506969 on Build GRCm38
- UniGene ID
- Mm.42234
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001162521.1 |
NP_001155993.1
|
3, 4 |
531 |
|
NM_013764.2 |
NP_038792.2
|
3, 4 |
531 |
|
GenBank mRNA |
AJ133749.1 |
- |
3, 4 |
456 |
|
AK035281.1 |
- |
3, 4 |
477 |
|
AK167071.1 |
- |
2, 3 |
292 |
|
AK185573.1 |
- |
1, 2 |
181 |
|
AY037862.1 |
- |
3, 4 |
461 |
|
BC012647.1 |
- |
3, 4 |
476 |
|
BC094920.1 |
- |
3, 4 |
531 |
30055743 |
BQ746075.1 |
- |
3, 4 |
445 |
|
U90524.1 |
- |
3, 4 |
451 |
|
View Full Product Details