Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178601.3 | NP_848716.2 | 7 | 704 | |
GenBank mRNA | AF334609.1 | - | 6 | 608 | |
AK004135.1 | - | 6 | 606 | ||
AK008178.1 | - | 7 | 666 | ||
AK010504.1 | - | 7 | 664 | ||
AK013334.1 | - | 7 | 633 | ||
AK018039.1 | - | 7 | 661 | ||
AK084933.1 | - | 7 | 640 | ||
AK151176.1 | - | 7 | 683 | ||
AK152214.1 | - | 7 | 683 | ||
AK166903.1 | - | 7 | 654 | ||
BC037602.1 | - | 6 | 625 | ||
BC069930.1 | - | 6 | 614 | 3481801 |