Gene Transcripts
- Gene Symbol
- Smtn
- Entrez Gene ID
- 29856
- Gene Name
- smoothelin
- Gene Aliases
- smsmo
- Chromosome Location
- Chr. 11: 3517522 - 3539654 on Build GRCm38
- UniGene ID
- Mm.188516
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001159284.1 |
NP_001152756.1
|
13, 14 |
2085 |
|
NM_001284427.1 |
NP_001271356.1
|
12, 13 |
2034 |
|
NM_001284428.1 |
NP_001271357.1
|
13, 14 |
2142 |
|
NM_001284429.1 |
NP_001271358.1
|
13, 14 |
2087 |
|
NM_013870.3 |
NP_038898.2
|
12, 13 |
2034 |
|
GenBank mRNA |
AF064236.1 |
- |
12, 13 |
1981 |
|
AJ010305.1 |
- |
12, 13 |
1981 |
|
AK009426.1 |
- |
2, 3 |
675 |
|
AK030932.1 |
- |
12, 13 |
2035 |
|
AK083947.1 |
- |
2, 3 |
288 |
|
AK085412.1 |
- |
13, 14 |
2049 |
|
AK138898.1 |
- |
13, 14 |
2100 |
|
AK160492.1 |
- |
3, 4 |
489 |
|
AK166385.1 |
- |
13, 14 |
2102 |
|
AK170516.1 |
- |
12, 13 |
1994 |
|
BC010599.1 |
- |
12, 13 |
1975 |
3709078 |
BC066192.1 |
- |
13, 14 |
1960 |
30433616 |
BC069836.1 |
- |
13, 14 |
1960 |
30456568 |
BC069839.1 |
- |
13, 14 |
1960 |
30431675 |
View Full Product Details