Gene Transcripts

Gene Symbol
Fbxw2
Entrez Gene ID
30050
Gene Name
F-box and WD-40 domain protein 2
Gene Aliases
2700071L08Rik, FBW2, Fwd2, MD6
Chromosome Location
Chr. 2: 34804364 - 34826273 on Build GRCm38
UniGene ID
Mm.4465
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164768.1 NP_001158240.1 1 397
NM_001164769.1 NP_001158241.1 3 456
NM_001164770.1 NP_001158242.1 3 364
NM_001164772.1 NP_001158244.1 3 456
NM_013890.4 NP_038918.3 3 456
GenBank mRNA AF140683.1 - 1 401
AK004015.1 - 3 422
AK005103.1 - 3 421
AK009893.1 - 2 478
AK014858.1 - 3 441
AK033129.1 - 3 410
AK040764.1 - 3 423
AK078815.1 - 3 364
AK088898.1 - 1 136
AK133758.1 - 1 3424
AK145405.1 - 3 423
AK184341.1 - 1 104
BC003834.1 - 3 542 3601733

View Full Product Details