Gene Transcripts

Gene Symbol
Fbxw2
Entrez Gene ID
30050
Gene Name
F-box and WD-40 domain protein 2
Gene Aliases
2700071L08Rik, FBW2, Fwd2, MD6
Chromosome Location
Chr. 2: 34804364 - 34826273 on Build GRCm38
UniGene ID
Mm.4465
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164768.1 NP_001158240.1 2 655
NM_001164769.1 NP_001158241.1 4 714
NM_001164770.1 NP_001158242.1 4 622
NM_001164772.1 NP_001158244.1 4 714
NM_013890.4 NP_038918.3 4 714
GenBank mRNA AF140683.1 - 2 659
AK004015.1 - 4 680
AK005103.1 - 4 679
AK009893.1 - 3 736
AK033129.1 - 4 668
AK040764.1 - 4 681
AK078815.1 - 4 622
AK088898.1 - 2 394
AK145405.1 - 4 681
AK179719.1 - 2 208
AK184341.1 - 2 362
BC003834.1 - 4 800 3601733
X54352.1 - 3 644

View Full Product Details