Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016964.2 | NP_058660.2 | 15 | 1689 | |
GenBank mRNA | AJ005678.1 | - | 15 | 1647 | |
AK145898.1 | - | 14 | 2176 | ||
BC060184.1 | - | 14 | 2130 | ||
BC139010.1 | - | 15 | 1619 | 8862032 | |
BC139011.1 | - | 15 | 1619 | 8862033 |