Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199020.1 | NP_001185949.1 | 3 | 425 | |
NM_021421.4 | NP_067396.3 | 2 | 560 | ||
GenBank mRNA | AK012003.1 | - | 2 | 556 | |
AK017545.1 | - | 2 | 523 | ||
AK077039.1 | - | 2 | 506 | ||
AK135709.1 | - | 3 | 422 | ||
AK158731.1 | - | 2 | 576 | ||
AK166619.1 | - | 3 | 424 | ||
BC024907.1 | - | 2 | 520 | 2812124 |