Gene Transcripts

Gene Symbol
Batf
Entrez Gene ID
53314
Gene Name
basic leucine zipper transcription factor, ATF-like
Gene Aliases
B-ATF, SFA-2
Chromosome Location
Chr. 12: 85686720 - 85709087 on Build GRCm38
UniGene ID
Mm.6672
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016767.2 NP_058047.1 3 411
GenBank mRNA AF017021.1 - 3 411
AK018587.1 - 3 453
BC132410.1 - 3 245 40130687
BC132412.1 - 3 245 40130689

View Full Product Details