Gene Transcripts
- Gene Symbol
- KPNA1
- Entrez Gene ID
- 3836
- Gene Name
- karyopherin subunit alpha 1
- Gene Aliases
- IPOA5, NPI-1, RCH2, SRP1
- Chromosome Location
- Chr. 3: 122421901 - 122514939 on Build GRCh38
- UniGene ID
- Hs.161008
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002264.3 |
NP_002255.3
|
2, 3 |
320 |
|
NR_026698.1 |
|
2, 3 |
320 |
|
XM_005247437.3 |
XP_005247494.1
|
2, 3 |
202 |
|
XM_011512797.2 |
XP_011511099.1
|
1, 2 |
137 |
|
XM_017006363.1 |
XP_016861852.1
|
2, 3 |
318 |
|
XM_017006364.1 |
XP_016861853.1
|
2, 3 |
197 |
|
GenBank mRNA |
AK299030.1 |
- |
1, 2 |
206 |
|
AK312713.1 |
- |
2, 3 |
315 |
|
BC002374.2 |
- |
2, 3 |
309 |
2822859 |
BC003009.1 |
- |
2, 3 |
309 |
2822859 |
BC090864.1 |
- |
2, 3 |
291 |
6173605 |
BT006959.1 |
- |
1, 2 |
132 |
|
CR456743.1 |
- |
1, 2 |
132 |
|
DA308391.1 |
- |
2, 3 |
320 |
|
DN991078.1 |
- |
2, 3 |
162 |
|
S75295.1 |
- |
2, 3 |
178 |
|
View Full Product Details