Gene Transcripts

Gene Symbol
Gpatch11
Entrez Gene ID
53951
Gene Name
G patch domain containing 11
Gene Aliases
2310002B06Rik, C80922, Ccdc75, L26697
Chromosome Location
Chr. 17: 78835516 - 78848308 on Build GRCm38
UniGene ID
Mm.220253
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181649.6 NP_857632.4 6 654
GenBank mRNA AK009084.1 - 6 541
BC092522.1 - 6 581 30648550

View Full Product Details