Gene Transcripts

Gene Symbol
KRT6A
Entrez Gene ID
3853
Gene Name
keratin 6A
Gene Aliases
CK-6C, CK-6E, CK6A, CK6C, CK6D, K6A, K6C, K6D, KRT6C, KRT6D, PC3
Chromosome Location
Chr. 12: 52487174 - 52493397 on Build GRCh38
UniGene ID
Hs.700779
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005554.3 NP_005545.1 9 2377
GenBank mRNA BC008807.2 - 9 2216
BC014152.2 - 9 2220 3639270
BC069269.1 - 9 2198 5428084
BG059253.1 - 1 60

View Full Product Details