Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001025600.1 | NP_001020771.1 | 3 | 344 | |
NM_018770.3 | NP_061240.3 | 3 | 344 | ||
NM_207675.2 | NP_997558.2 | 3 | 344 | ||
NM_207676.2 | NP_997559.1 | 3 | 344 | ||
GenBank mRNA | AB052293.1 | - | 3 | 344 | |
AB064265.2 | - | 3 | 323 | ||
AB092414.1 | - | 3 | 323 | ||
AB183399.1 | - | 3 | 323 | ||
AB183400.1 | - | 3 | 323 | ||
AB183401.1 | - | 3 | 323 | ||
AB183402.1 | - | 3 | 323 | ||
AB377192.1 | - | 3 | 344 | ||
AF061260.1 | - | 2 | 181 | ||
AF434663.1 | - | 3 | 439 | ||
AF539424.1 | - | 3 | 336 | ||
AK013775.1 | - | 3 | 345 | ||
AK013911.1 | - | 3 | 438 | ||
AK053839.1 | - | 3 | 345 | ||
AK172628.1 | - | 2 | 1936 | ||
AY351388.1 | - | 3 | 323 | ||
BC057125.1 | - | 3 | 332 | 6414884 | |
BC090970.1 | - | 3 | 523 | 6703838 | |
BC095986.1 | - | 3 | 336 | 5372277 | |
DQ279856.1 | - | 3 | 323 | ||
JQ782219.1 | - | 3 | 323 |