Gene Transcripts

Gene Symbol
Nme4
Entrez Gene ID
56520
Gene Name
NME/NM23 nucleoside diphosphate kinase 4
Gene Aliases
2610027N22Rik, 2810024O08Rik, 5730493H09Rik, NM23-M4, Nm23M4
Chromosome Location
Chr. 17: 26091734 - 26095527 on Build GRCm38
UniGene ID
Mm.41787
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019731.1 NP_062705.1 5 741
GenBank mRNA AF153451.1 - 5 741
AF288690.1 - 5 754
AK012805.1 - 5 777
AK017724.1 - 5 757
BC027277.1 - 5 759 3591383

View Full Product Details