Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252463.1 | NP_001239392.1 | 7 | 690 | |
NM_001252464.1 | NP_001239393.1 | 7 | 665 | ||
NM_001252465.1 | NP_001239394.1 | 5 | 474 | ||
NM_019988.5 | NP_064372.2 | 7 | 685 | ||
GenBank mRNA | AK002751.1 | - | 7 | 651 | |
AK038515.1 | - | 6 | 1183 | ||
AK040100.1 | - | 7 | 686 | ||
AK048102.1 | - | 7 | 649 | ||
AK077680.1 | - | 7 | 656 | ||
AK083731.1 | - | 7 | 626 | ||
AK088951.1 | - | 7 | 659 | ||
BC015279.1 | - | 7 | 644 | 4222934 | |
BM947104.1 | - | 5 | 456 | ||
CF750475.1 | - | 7 | 654 |