Gene Transcripts

Gene Symbol
Clec1b
Entrez Gene ID
56760
Gene Name
C-type lectin domain family 1, member b
Gene Aliases
1810061I13Rik, Clec-2, Clec2
Chromosome Location
Chr. 6: 129397297 - 129405413 on Build GRCm38
UniGene ID
Mm.30700
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204239.1 NP_001191168.1 3 583
NM_019985.3 NP_064369.1 4 679
GenBank mRNA AF201457.1 - 4 390
AK007925.1 - 4 659
BC064054.1 - 4 679 574821
EF070191.1 - 3 273

View Full Product Details