Gene Transcripts

Gene Symbol
Akip1
Entrez Gene ID
57373
Gene Name
A kinase (PRKA) interacting protein 1
Gene Aliases
BCA3, C11orf17, D7H11orf17, D930014E17Rik, ICRFP703B1614Q5.6, ORF27
Chromosome Location
Chr. 7: 109703670 - 109712186 on Build GRCm38
UniGene ID
Mm.334138
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020616.1 NP_065641.1 6 593
GenBank mRNA AK086219.1 - 6 626
AK155010.1 - 6 642
AK159903.1 - 6 594
AK163012.1 - 6 640

View Full Product Details