Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020590.4 | NP_065615.1 | 1 | 296 | |
GenBank mRNA | AB041648.1 | - | 1 | 320 | |
AF180518.1 | - | 1 | 323 | ||
AK006103.1 | - | 1 | 133 | ||
AK011975.1 | - | 1 | 291 | ||
AK014083.1 | - | 1 | 110 | ||
AK015049.1 | - | 1 | 113 | ||
AK018687.1 | - | 1 | 297 | ||
AK159015.1 | - | 1 | 291 | ||
AK167500.1 | - | 1 | 289 | ||
AK168921.1 | - | 1 | 328 | ||
AK182413.1 | - | 1 | 70 | ||
BC004602.1 | - | 1 | 231 | 3585132 | |
BC024706.1 | - | 1 | 295 | 3711973 |