Gene Transcripts
- Gene Symbol
- Fbxo3
- Entrez Gene ID
- 57443
- Gene Name
- F-box protein 3
- Gene Aliases
- 1200002G09Rik, 1700026K02Rik, AI046358, Fba
- Chromosome Location
- Chr. 2: 104027799 - 104063237 on Build GRCm38
- UniGene ID
- Mm.143768
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020593.2 |
NP_065618.1
|
1, 2 |
168 |
|
NM_212433.1 |
NP_997598.1
|
1, 2 |
168 |
|
GenBank mRNA |
AF233226.1 |
- |
1, 2 |
107 |
|
AK004544.2 |
- |
1, 2 |
120 |
|
AK009024.1 |
- |
1, 2 |
108 |
|
AK019550.1 |
- |
1, 2 |
108 |
|
AK028048.1 |
- |
1, 2 |
130 |
|
AK031185.1 |
- |
1, 2 |
107 |
|
AK031428.1 |
- |
1, 2 |
128 |
|
AK049870.1 |
- |
1, 2 |
126 |
|
AK050195.1 |
- |
1, 2 |
120 |
|
AK085686.1 |
- |
1, 2 |
123 |
|
AK150097.1 |
- |
1, 2 |
120 |
|
AK151130.1 |
- |
1, 2 |
119 |
|
AK151857.1 |
- |
1, 2 |
119 |
|
AK152780.1 |
- |
1, 2 |
119 |
|
AK167075.1 |
- |
1, 2 |
106 |
|
AK215292.1 |
- |
1, 2 |
112 |
|
AK215501.1 |
- |
1, 2 |
112 |
|
BC010212.1 |
- |
1, 2 |
69 |
|
BC050758.1 |
- |
1, 2 |
121 |
|
BC058253.1 |
- |
1, 2 |
125 |
6465107 |
BY105498.1 |
- |
1, 2 |
168 |
|
View Full Product Details