Gene Transcripts

Gene Symbol
Crlf2
Entrez Gene ID
57914
Gene Name
cytokine receptor-like factor 2
Gene Aliases
CRLM2, Ly114, Tpte2, Tslpr
Chromosome Location
Chr. 5: 109554709 - 109559017 on Build GRCm38
UniGene ID
Mm.35771
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164735.1 NP_001158207.1 4 470
NM_016715.4 NP_057924.3 4 470
GenBank mRNA AB031333.1 - 4 418
AB039945.1 - 4 366
AF201963.1 - 4 521
AF232936.1 - 4 434
AK149928.1 - 4 471
AK170466.1 - 1 332
BC016580.1 - 4 548 2655857
BC023788.1 - 4 458 5346748

View Full Product Details