Gene Transcripts

Gene Symbol
Shank3
Entrez Gene ID
58234
Gene Name
SH3/ankyrin domain gene 3
Gene Aliases
AI841104, SHANK3c-3, SHANK3c-4, Shank3b
Chromosome Location
Chr. 15: 89499707 - 89560261 on Build GRCm38
UniGene ID
Mm.146855
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021423.3 NP_067398.2 3 279
GenBank mRNA AB231013.1 - 3 279
AJ245904.1 - 1 133

View Full Product Details