Gene Transcripts

Gene Symbol
Pga5
Entrez Gene ID
58803
Gene Name
pepsinogen 5, group I
Gene Aliases
1110035E17Rik, Pepf
Chromosome Location
Chr. 19: 10668957 - 10678071 on Build GRCm38
UniGene ID
Mm.34037
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021453.4 NP_067428.2 7 965
GenBank mRNA AF240776.1 - 7 874
AK004109.1 - 7 904
AK053965.1 - 7 966
AK168924.1 - 7 904
BC119523.2 - 7 923 40056066

View Full Product Details