Gene Transcripts

Gene Symbol
Myot
Entrez Gene ID
58916
Gene Name
myotilin
Gene Aliases
5530402I04Rik, Ttid
Chromosome Location
Chr. 18: 44334053 - 44355740 on Build GRCm38
UniGene ID
Mm.143804
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033621.3 NP_001028793.1 9 1541
GenBank mRNA AF230979.2 - 9 1540
AK017447.1 - 9 1542
BC016214.1 - 9 1519 4482382
BC064052.1 - 9 1514 1366418

View Full Product Details