Gene Transcripts

Gene Symbol
Moap1
Entrez Gene ID
64113
Gene Name
modulator of apoptosis 1
Gene Aliases
AA987038, Map-1, Pnma4
Chromosome Location
Chr. 12: 102739830 - 102743661 on Build GRCm38
UniGene ID
Mm.291222
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142937.2 NP_001136409.1 2 473
NM_022323.7 NP_071718.1 2 517
GenBank mRNA AF305551.1 - 1 248
AU080670.1 - 2 457
BU614788.1 - 2 487
BY741099.1 - 2 471

Gene Symbol
Gm20604
Entrez Gene ID
100859931
Gene Name
predicted gene 20604
Gene Aliases
AK010878-Moap1
Chromosome Location
Chr. 12: 102739830 - 102757810 on Build GRCm38
UniGene ID
Mm.291222
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142939.1 NP_001136411.1 2 624
GenBank mRNA AK011927.1 - 2 609
AK019599.1 - 1487
BC014715.1 - 2 573 4236212
BC055374.1 - 2 577 30071431

View Full Product Details