Gene Transcripts

Gene Symbol
Lpin2
Entrez Gene ID
64898
Gene Name
lipin 2
Gene Aliases
2610511G02Rik, AI481352, AW742896
Chromosome Location
Chr. 17: 71183528 - 71249818 on Build GRCm38
UniGene ID
Mm.227924
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164885.1 NP_001158357.1 8, 9 1495
NM_022882.4 NP_075020.2 8, 9 1371
GenBank mRNA AF286723.1 - 8, 9 1371
BC039698.1 - 8, 9 1484 5101211

View Full Product Details