Gene Transcripts

Gene Symbol
PRICKLE3
Entrez Gene ID
4007
Gene Name
prickle planar cell polarity protein 3
Gene Aliases
LMO6
Chromosome Location
Chr. X: 49174802 - 49186400 on Build GRCh38
UniGene ID
Hs.732712
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001307979.1 NP_001294908.1 3, 4 338
NM_006150.4 NP_006141.2 3, 4 415
GenBank mRNA AJ011654.1 - 3, 4 338
AK303200.1 - 3, 4 321
AK314788.1 - 3, 4 366
BC002468.2 - 3, 4 368 3347651
BC016856.1 - 3, 4 384 3846372
BP227957.1 - 3, 4 403
BT007423.1 - 3, 4 312
CR457211.1 - 3, 4 312

View Full Product Details