Gene Transcripts

Gene Symbol
Ska2
Entrez Gene ID
66140
Gene Name
spindle and kinetochore associated complex subunit 2
Gene Aliases
1110001A07Rik, C78640, Fam33a
Chromosome Location
Chr. 11: 87109261 - 87122974 on Build GRCm38
UniGene ID
Mm.45008
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025377.3 NP_079653.1 3 214
GenBank mRNA AK003196.1 - 3 242
AK013317.1 - 3 215
AK013370.1 - 3 215
AK017811.1 - 3 215
AK153219.1 - 185
AK162628.1 - 211
BC052151.1 - 215
BC060088.1 - 217
BC096627.1 - 226
BC117001.1 - 3 244 40126320
BC119167.1 - 3 244 8733916

View Full Product Details