Gene Transcripts
- Gene Symbol
- Mrpl4
- Entrez Gene ID
- 66163
- Gene Name
- mitochondrial ribosomal protein L4
- Gene Aliases
- 1110017G11Rik
- Chromosome Location
- Chr. 9: 21002605 - 21008839 on Build GRCm38
- UniGene ID
- Mm.427576
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023167.2 |
NP_075656.2
|
4, 5 |
532 |
|
GenBank mRNA |
AB041599.1 |
- |
3, 4 |
283 |
|
AB049634.1 |
- |
4, 5 |
531 |
|
AK002464.1 |
- |
4, 5 |
368 |
|
AK003746.2 |
- |
4, 5 |
489 |
|
BC023078.1 |
- |
4, 5 |
313 |
|
BC037064.1 |
- |
4, 5 |
365 |
|
BC039983.1 |
- |
4, 5 |
500 |
4952757 |
BC051685.1 |
- |
4, 5 |
345 |
|
BC061095.1 |
- |
4, 5 |
348 |
6588836 |
View Full Product Details