Gene Transcripts

Gene Symbol
Aqp11
Entrez Gene ID
66333
Gene Name
aquaporin 11
Gene Aliases
1700015P13Rik, AI648923, sjds
Chromosome Location
Chr. 7: 97725490 - 97738514 on Build GRCm38
UniGene ID
Mm.29756
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175105.3 NP_780314.1 2 956
GenBank mRNA AB028148.1 - 2 710
AK006005.1 - 2 981
AK075757.1 - 2 957
AK137326.1 - 2 1000
BC037088.1 - 2 735
BC068103.1 - 3 605

View Full Product Details