Gene Transcripts

Gene Symbol
Blzf1
Entrez Gene ID
66352
Gene Name
basic leucine zipper nuclear factor 1
Gene Aliases
1700030G05Rik, Blzf1l, Golgin-45, Jem-1
Chromosome Location
Chr. 1: 164289800 - 164307484 on Build GRCm38
UniGene ID
Mm.33285
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160208.1 NP_001153680.1 4, 5 760
NM_001160209.1 NP_001153681.1 3, 4 847
NM_025505.4 NP_079781.2 5, 6 781
GenBank mRNA AK005489.1 - 3, 4 581
AK006544.1 - 4, 5 764
AK029787.1 - 5, 6 782
AK167493.1 - 3, 4 854
AK168933.1 - 3, 4 608
AK171167.1 - 3, 4 639
BC027025.1 - 3, 4 622 5374749

View Full Product Details