Gene Transcripts

Gene Symbol
Cul7
Entrez Gene ID
66515
Gene Name
cullin 7
Gene Aliases
2510004L20Rik, AA409809, C230011P08Rik, p185, p193
Chromosome Location
Chr. 17: 46650338 - 46664364 on Build GRCm38
UniGene ID
Mm.329078
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025611.5 NP_079887.3 13 3066
GenBank mRNA AK147156.1 - 13 2824
AK147905.1 - 13 2785
AK155583.1 - 13 3075
AK170580.1 - 13 3070
AK170808.1 - 13 2840
AK172882.1 - 12 2139
BC019645.1 - 3 354
BC065096.1 - 12 2553
BC138998.1 - 13 3004 8862020
BC139000.1 - 13 3004 8862022

View Full Product Details