Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025725.2 | NP_080001.1 | 1 | 1429 | |
GenBank mRNA | AK014878.1 | - | 1 | 1427 | |
AK015646.1 | - | 1 | 1427 | ||
AK030023.1 | - | 1 | 1429 | ||
BC125034.1 | - | 1 | 1413 | 40096237 |