Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011416.2 | NP_035546.2 | 29, 30 | 4457 | |
NM_026003.2 | NP_080279.1 | 5, 6 | 606 | ||
GenBank mRNA | AK009309.1 | - | 1, 2 | 116 | |
AK011935.1 | - | 5, 6 | 551 | ||
AK147322.1 | - | 29, 30 | 4606 | ||
BC004653.1 | - | 1, 2 | 104 | ||
BC022975.1 | - | 7, 8 | 1083 | ||
BC056199.1 | - | 19, 20 | 2555 | ||
BC075641.1 | - | 29, 30 | 4439 | 30544092 |