Gene Transcripts

Gene Symbol
Armcx2
Entrez Gene ID
67416
Gene Name
armadillo repeat containing, X-linked 2
Gene Aliases
3230401N03Rik, AI043003, ALEX2
Chromosome Location
Chr. X: 134804142 - 134809221 on Build GRCm38
UniGene ID
Mm.285969
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166397.1 NP_001159869.1 6 2247
NM_001166398.1 NP_001159870.1 6 2190
NM_026139.4 NP_080415.3 5 2354
GenBank mRNA AK014329.1 - 6 2023
AK034185.1 - 6 2023
AK034621.1 - 6 2021
AK043440.1 - 6 2023
AK044926.1 - 6 2021
AK053354.1 - 5 2128
AK077990.1 - 1 1132
AK081299.1 - 6 1966
AK172960.1 - 6 2247
BC049088.1 - 6 2002
BC052383.1 - 6 1996
BC054839.1 - 3 1586 6490133

View Full Product Details