Gene Transcripts

Gene Symbol
Samd8
Entrez Gene ID
67630
Gene Name
sterile alpha motif domain containing 8
Gene Aliases
1110053F04Rik, 1700010P07Rik, SMSr
Chromosome Location
Chr. 14: 21750531 - 21798725 on Build GRCm38
UniGene ID
Mm.102765
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026283.2 NP_080559.1 2 770
GenBank mRNA AK006211.1 - 2 771
AK015593.1 - 2 748
AK075749.1 - 2 721
AK147234.1 - 2 574
AK147528.1 - 2 574
BC100476.1 - 2 785 30916247
BC116745.1 - 2 758 40126064
BC116747.1 - 2 758 40126066

View Full Product Details