Gene Transcripts

Gene Symbol
Rbm18
Entrez Gene ID
67889
Gene Name
RNA binding motif protein 18
Gene Aliases
2010004P11Rik, AU015530
Chromosome Location
Chr. 2: 36116079 - 36136704 on Build GRCm38
UniGene ID
Mm.205937
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159635.1 NP_001153107.1 2 314
NM_026434.4 NP_080710.1 2 314
GenBank mRNA AK008110.1 - 2 170
AK021243.1 - 2 195
AK035348.1 - 2 222
AK036220.1 - 1 2440
AK036252.1 - 2 195
AK135109.1 - 2 222
BB841783.1 - 2 223
BC005540.1 - 2 208 3590164
BY009724.1 - 2 221
CB183820.1 - 2 385

View Full Product Details