Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026558.4 | NP_080834.3 | 7, 8 | 949 | |
GenBank mRNA | AK013129.1 | - | 6, 7 | 778 | |
AK019574.1 | - | 7, 8 | 777 | ||
AK078438.1 | - | 7, 8 | 930 | ||
AK088705.1 | - | 7, 8 | 930 | ||
AK214863.1 | - | 1, 2 | 24 | ||
BC020162.1 | - | 743 | 3991990 | ||
BC055848.1 | - | 7, 8 | 1867 |