Gene Transcripts

Gene Symbol
Wfdc15a
Entrez Gene ID
68221
Gene Name
WAP four-disulfide core domain 15A
Gene Aliases
1700049M11Rik, Wfdcl1
Chromosome Location
Chr. 2: 164198872 - 164200117 on Build GRCm38
UniGene ID
Mm.142735
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183271.2 NP_899094.1 1, 2 166
GenBank mRNA AK006775.1 - 1, 2 160
AK018854.1 - 1, 2 160
AK018888.1 - 1, 2 167
AK018909.1 - 1, 2 138
BC100423.1 - 1, 2 145 30919644

View Full Product Details