Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_197982.3 | NP_932099.2 | 10 | 1330 | |
GenBank mRNA | AK011285.1 | - | 9 | 1169 | |
AK012002.1 | - | 10 | 1237 | ||
AK016960.1 | - | 10 | 1239 | ||
AK088894.1 | - | 4 | 632 | ||
AK137114.1 | - | 1236 | |||
AK145927.1 | - | 10 | 1273 | ||
AK146294.1 | - | 10 | 1275 | ||
AK149475.1 | - | 10 | 1672 | ||
AK150997.1 | - | 10 | 1259 | ||
AK151395.1 | - | 11 | 1299 | ||
AK152394.1 | - | 10 | 1240 | ||
AK162752.1 | - | 1798 | |||
AK164289.1 | - | 10 | 1200 | ||
AK167890.1 | - | 10 | 1200 | ||
AK168079.1 | - | 10 | 1199 | ||
AK172448.1 | - | 10 | 1259 | ||
AK195258.1 | - | 4 | 320 | ||
BC020134.1 | - | 10 | 1218 | 3969048 |